missing translation for 'onlineSavingsMsg'
Learn More

4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15439395
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448637-05-5
Molecular Formula C22H23BO4
Molecular Weight (g/mol) 362.23
MDL Number MFCD16294527
InChI Key SGGJQXOMPWFXNR-UHFFFAOYSA-N
Synonym methyl 4-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-benzoic acid methyl ester, amtb207, 4-4-methoxycarbonyl phenyl ethynyl benzeneboronic acid pinacol ester, methyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl benzoic acid methyl ester
PubChem CID 71306515
IUPAC Name methyl 4-[2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 362.23
Percent Purity 95%
Chemical Name or Material 4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'