missing translation for 'onlineSavingsMsg'
Learn More
{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™ Product Code.: 10778645

{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™

Product Code. 10778645
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10778645

Brand: Thermo Scientific Maybridge CC55809DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-50-0
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
MDL Number MFCD09064973
InChI Key YQUWAYPNHCWYNU-UHFFFAOYSA-N
Synonym 4-4-methylperhydro-1,4-diazepin-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazepan-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazaperhydroepinyl methyl phenyl methan-1-ol, benzenemethanol,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229552
IUPAC Name [4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl]methanol
SMILES CN1CCCN(CC1)CC2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material {4-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]phenyl}methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.