missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, 97%, Thermo Scientific™

Product Code. p-4509924
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10381293

Brand: Thermo Scientific Maybridge CC45604CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 439691-80-2
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
InChI Key DJJFKXUSAXIMLS-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzaldehyde, 1-4-formylbenzyl-4-methylpiperazine, benzaldehyde,4-4-methyl-1-piperazinyl methyl, 4-4-methylpiperazino methyl benzaldehyde, 4-4-methylpiperazinyl methyl benzaldehyde, 4-4-methyl piperazin-1-ylmethyl benzaldehyde, 4-4-methyl-1-piperazinyl methyl benzaldehyde
PubChem CID 7164651
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 218.3
Percent Purity 97%
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.