missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Trifluoromethoxy)phenoxy]piperidine, 97%, Thermo Scientific™

Product Code. 11414810
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287952-67-4
Molecular Formula C12H14F3NO2
Molecular Weight (g/mol) 261.24
MDL Number MFCD06656166
InChI Key RPQOTFPZKNHYFB-UHFFFAOYSA-N
Synonym 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine
PubChem CID 16115439
IUPAC Name 4-[4-(trifluoromethoxy)phenoxy]piperidine
SMILES FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1

Specifications

Melting Point 71°C to 73°C
Quantity 250 mg
Formula Weight 261.25
Percent Purity 97%
Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'