missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4',5,7-Trihydroxyisoflavone, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
10 g
Description
4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Solubility
Soluble in chloroform, methanol. Insoluble in cold water.
Notes
Protect from heat. Store away from oxidizing agents.
4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Solubility
Soluble in chloroform, methanol. Insoluble in cold water.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | ∼300°C |
Color | Brown to White |
Quantity | 10 g |
Beilstein | 263823 |
Merck Index | 14,4391 |
Solubility Information | Soluble in chloroform,methanol. Insoluble in cold water. |
Formula Weight | 270.24 |
Percent Purity | ≥99% |
Physical Form | Powder |
Chemical Name or Material | 4',5,7-Trihydroxyisoflavone |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction