missing translation for 'onlineSavingsMsg'
Learn More
4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%, Thermo Scientific Chemicals Product Code.: 11336699

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%, Thermo Scientific Chemicals

Product Code. 11336699
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11336699

Brand: Thermo Scientific Alfa Aesar H26241.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

Solubility
Soluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161265-03-8
Molecular Formula C39H32OP2
Molecular Weight (g/mol) 578.63
MDL Number MFCD00233866
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xantheneShow MoreShow Less
PubChem CID 636044
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 228°C
Quantity 1 g
Solubility Information Soluble in organic solvents.
Formula Weight 578.62
Percent Purity 97%
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX05C011 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH26241.06,H26241.03,H26241.14
Lot NumberU02H096 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH26241.06,H26241.03,H26241.14
Lot NumberP31C014 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH26241.06,H26241.03,H26241.14
Lot NumberL22T016 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26241.06,H26241.03,H26241.14
Lot NumberD17T029 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26241.06,H26241.03,H26241.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.