missing translation for 'onlineSavingsMsg'
Learn More

4-[(5-Bromo-2-furyl)methyl]morpholine, 97%, Thermo Scientific™

Product Code. 10366324
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 71897-83-1
Molecular Formula C9H12BrNO2
Molecular Weight (g/mol) 246.104
MDL Number MFCD01307635
InChI Key KNWZHIWPPIXAJQ-UHFFFAOYSA-N
Synonym 4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl
PubChem CID 12478687
IUPAC Name 4-[(5-bromofuran-2-yl)methyl]morpholine
SMILES C1COCCN1CC2=CC=C(O2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 246.1
Percent Purity 97%
Chemical Name or Material 4-[(5-Bromo-2-furyl)methyl]morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'