missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromo-2-thienyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Product Code. 10019081
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 34801-14-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5BrN2S2
MDL Number MFCD01812622
Quantity 1 g
Synonym 4-5-bromothiophen-2-yl thiazol-2-amine, 4-5-bromothiophen-2-yl-1,3-thiazol-2-amine, 4-5-bromothiophen-2-yl thiazol-2-ylamine, 4-5-bromo-2-thienyl-1,3-thiazol-2-amine, 4-5-bromo-thiophen-2-yl-thiazol-2-ylamine, 2-amino-4-5-bromothien-2-yl thiazole, 4-5-bromothien-2-yl-1,3-thiazol-2-amine, 4-5-bromo-2-thienyl-1,3-thiazole-2-ylamine, 2-thiazolamine, 4-5-bromo-2-thienyl, maybridge1_001930
InChI Key MMZHPDCFNLBMBY-UHFFFAOYSA-N
SMILES C1=C(SC(=C1)Br)C2=CSC(=N2)N
IUPAC Name 4-(5-bromothiophen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol) 261.155
PubChem CID 725239
Formula Weight 261.17
Percent Purity 97%
Chemical Name or Material 4-(5-Bromo-2-thienyl)-1, 3-thiazol-2-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(5-Bromo-2-thienyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.