missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 4',5'-Dibromofluorescein, ca. 95% dye content Product Code.: 11345159

Thermo Scientific Chemicals 4',5'-Dibromofluorescein, ca. 95% dye content

Product Code. 11345159
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11345159

Brand: Thermo Scientific Chemicals A18226.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-03-2
Molecular Formula C20H10Br2O5
Molecular Weight (g/mol) 490.103
MDL Number MFCD00005042
InChI Key ZDTNHRWWURISAA-UHFFFAOYSA-N
Synonym 4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201
PubChem CID 11689
IUPAC Name 4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O

Specifications

Melting Point 270°C to 273°C
Quantity 50 g
Beilstein 57189
Merck Index 14,3022
Formula Weight 490.12
Percent Purity 95%
Chemical Name or Material 4',5'-Dibromofluorescein, ≈95% dye content

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4',5'-Dibromofluorescein, ca. 95% dye content >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.