missing translation for 'onlineSavingsMsg'
Learn More
4',5'-Dibromofluorescein, Thermo Scientific™ Product Code.: 15452132

4',5'-Dibromofluorescein, Thermo Scientific™

Product Code. 15452132
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15452132

Brand: Thermo Scientific Acros 202431000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 596-03-2
Molecular Formula C20H10Br2O5
Molecular Weight (g/mol) 490.11
InChI Key ZDTNHRWWURISAA-UHFFFAOYSA-N
Synonym 4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201
PubChem CID 11689
IUPAC Name 4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O

Specifications

Melting Point 270.0°C to 273.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 19,V,6,462
Merck Index 15,3027
Solubility Information (1% in hot 0.5 N NH4OH) Clear yellow to orange
Formula Weight 490.11
Chemical Name or Material 4',5'-Dibromofluorescein
compliance-icons
Product Identifier
  • 4',5'-Dibromofluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4',5'-Dibromofluorescein, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.