missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%

Product Code. 10011790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10011790

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, 98%, C10H6Na2O8S2.2H2O, CAS Number-5808-22-0 | CAS: 5808-22-0 | C10H6O8S2 | 318.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5808-22-0
Molecular Formula C10H10Na2O10S2
Molecular Weight (g/mol) 400.28
MDL Number MFCD00150612
InChI Key QUEAKWJKJBFNEG-UHFFFAOYSA-L
Synonym chromotropic acid sodium salt, 4,5-dihydroxy-2,7-naphthalenedisulfonate, di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid, 4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g
PubChem CID 124202444
IUPAC Name 2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
SMILES O.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300.0°C
Color Beige-Gray to White or Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 11, 307
Merck Index 15, 2245
Solubility Information Solubility in water: very soluble
Formula Weight 400.28
Percent Purity 98%
Insoluble Matter 0.02% max. (in water)
Physical Form Powder
Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.