missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dimethoxy-2-nitrobenzyl chloroformate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, acetone. Reacts with water.
Notes
Moisture and light sensitive. Store away from oxidizing agents, light, water/ moisture, heat, bases.
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, acetone. Reacts with water.
Notes
Moisture and light sensitive. Store away from oxidizing agents, light, water/ moisture, heat, bases.
Specifications
Specifications
Melting Point | ∽120°C (decomposition) |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 2389168 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in methanol,acetone. Reacts with water. |
Formula Weight | 275.64 |
Percent Purity | 97% |
Chemical Name or Material | 4,5-Dimethoxy-2-nitrobenzyl chloroformate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'