missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-o-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11358366
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2'-dithiobis(1-aminonaphthalene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).

Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131076-14-7
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.11
MDL Number MFCD00190674
InChI Key ORAAOAMEUMZGGU-UHFFFAOYSA-N
Synonym 4,5-dimethoxybenzene-1,2-diamine dihydrochloride, 4,5-dimethoxy-1,2-phenylenediamine dihydrochloride, 1,2-benzenediamine, 4,5-dimethoxy-, dihydrochloride, 4,5-dimethoxybenzene-1,2-bis aminium dichloride, acmc-1c0pe, 4,5-diaminoveratrol-dihydrochlorid, 4,5-dimethoxy-o-phenylenediamine dihydrochloride, 1,2-diamino-4,5-dimethoxybenzene hydrochloride, 4,5-dimethoxy-benzene-1,2-diamine dihydrochloride, 1,2-diamino-4,5-dimethoxybenzene, dihydrochloride
PubChem CID 13000219
SMILES Cl.Cl.COC1=C(OC)C=C(N)C(N)=C1

Specifications

Melting Point ∽265°C (decomposition)
Quantity 250 mg
UN Number UN2811
Beilstein 3714004
Solubility Information Soluble in warm methanol,dimethyl sulfoxide and water.
IUPAC Name 4,5-dimethoxybenzene-1,2-diamine;dihydrochloride
Formula Weight 241.12
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-o-phenylenediamine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'