missing translation for 'onlineSavingsMsg'
Learn More

4(5)-Imidazolemethanol hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11406900
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It has been used in the studies on some heterocyclic ligands showing linked ionization effects. 4-(Hydroxymethyl)imidazole HCl has been used in the synthesis of imidazole-containing peptidomimetic inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the studies on some heterocyclic ligands showing linked ionization effects. 4-(Hydroxymethyl)imidazole HCl has been used in the synthesis of imidazole-containing peptidomimetic inhibitors.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32673-41-9
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.56
MDL Number MFCD00012697
InChI Key WFNASTYGEKUMIY-UHFFFAOYSA-N
Synonym 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride
PubChem CID 122926
IUPAC Name 1H-imidazol-5-ylmethanol;hydrochloride
SMILES Cl.OCC1=CN=CN1

Specifications

Melting Point 104°C to 108°C
Quantity 5 g
Beilstein 3562041
Solubility Information Soluble in water.
Formula Weight 134.57
Percent Purity ≥98%
Chemical Name or Material 4(5)-Imidazolemethanol hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'