missing translation for 'onlineSavingsMsg'
Learn More

4(5)-Imidazolemethanol hydrochloride, 98+%

Product Code. p-7048064
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11406900

Brand: Thermo Scientific Alfa Aesar L20182.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32673-41-9 | C4H7ClN2O | 134.56 g/mol

It has been used in the studies on some heterocyclic ligands showing linked ionization effects. 4-(Hydroxymethyl)imidazole HCl has been used in the synthesis of imidazole-containing peptidomimetic inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the studies on some heterocyclic ligands showing linked ionization effects. 4-(Hydroxymethyl)imidazole HCl has been used in the synthesis of imidazole-containing peptidomimetic inhibitors.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32673-41-9
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.56
MDL Number MFCD00012697
InChI Key WFNASTYGEKUMIY-UHFFFAOYSA-N
Synonym 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride
PubChem CID 122926
IUPAC Name 1H-imidazol-5-ylmethanol;hydrochloride
SMILES Cl.OCC1=CN=CN1

Specifications

Melting Point 104°C to 108°C
Quantity 5 g
Beilstein 3562041
Solubility Information Soluble in water.
Formula Weight 134.57
Percent Purity ≥98%
Chemical Name or Material 4(5)-Imidazolemethanol hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.