missing translation for 'onlineSavingsMsg'
Learn More

4,6-Decadiyne, 97%, Thermo Scientific Chemicals

54.27 € - 691.44 €

Chemical Identifiers

CAS 16387-71-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00041645
InChI Key LIWZSNTUMSGWTF-UHFFFAOYSA-N
Synonym 4,6-decadiyne, acmc-1c48i
PubChem CID 140067
IUPAC Name deca-4,6-diyne
SMILES CCCC#CC#CCCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15480308
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53467.03
1 g
54.27 €
1g
Please to purchase this item. Need a web account? Register with us today!
15490308
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53467.06
5 g
177.89 €
5g
Please to purchase this item. Need a web account? Register with us today!
15400318
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53467.14
25 g
691.44 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

Solubility
Immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

16387-71-6
134.222
LIWZSNTUMSGWTF-UHFFFAOYSA-N
140067
CCCC#CC#CCCC
C10H14
MFCD00041645
4,6-decadiyne, acmc-1c48i
deca-4,6-diyne

Specifications

16387-71-6
96°C (15 mmHg)
CH3(CH2)2C≡CC≡C(CH2)2CH3
4,6-decadiyne, acmc-1c48i
LIWZSNTUMSGWTF-UHFFFAOYSA-N
deca-4,6-diyne
140067
97%
0.82
C10H14
MFCD00041645
Immiscible with water.
CCCC#CC#CCCC
134.222
134.22
4,6-Decadiyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only