missing translation for 'onlineSavingsMsg'
Learn More

4,6-Difluoroindole, 97%, Thermo Scientific Chemicals

37.60 € - 439.00 €

Chemical Identifiers

CAS 199526-97-1
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD01075213
InChI Key MHICCULQVCEWFH-UHFFFAOYSA-N
PubChem CID 2778736
IUPAC Name 4,6-difluoro-1H-indole
SMILES C1=CNC2=CC(=CC(=C21)F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410818
View Documents
Thermo Scientific Alfa Aesar
H54536.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15490808
View Documents
Thermo Scientific Alfa Aesar
H54536.03
1 g
113.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15400818
View Documents
Thermo Scientific Alfa Aesar
H54536.06
5 g
439.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.

Notes
Incompatible with oxidizing agents. Keep at room temperature.
Specifications

Chemical Identifiers

199526-97-1
153.132
MHICCULQVCEWFH-UHFFFAOYSA-N
4,6-difluoro-1H-indole
C8H5F2N
MFCD01075213
2778736
C1=CNC2=CC(=CC(=C21)F)F

Specifications

199526-97-1
MFCD01075213
4,6-difluoroindole, 1h-indole, 4,6-difluoro, 4,6-difluoro-indole, pubchem7179, acmc-209f4a, 1h-indole,4,6-difluoro, 1h-indole,4,6-difluoro-9ci, 4,6-difluoroindole 153.13, 4,6-difluoro-1h-indole; 1h-indole,4,6-difluoro-9ci
MHICCULQVCEWFH-UHFFFAOYSA-N
4,6-difluoro-1H-indole
2778736
97%
C8H5F2N
Air sensitive
Sparingly Soluble (0.062g/L) (25°C),Calc.
C1=CNC2=CC(=CC(=C21)F)F
153.132
153.13
4,6-Difluoroindole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.