missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol, 97%, Thermo Scientific™

Product Code. 11459926
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93892-40-1
Molecular Formula C19H30O
Molecular Weight (g/mol) 274.448
MDL Number MFCD00061045
InChI Key WGQDDNQFNLMBOS-UHFFFAOYSA-N
Synonym 4,6-diisopropyl-1,1,3,3-tetramethyl-5-indanol, 4,6-bis isopropyl-1,1,3,3-tetramethylindan-5-ol, 4,6-diisopropyl-1,1,3,3-tetramethyl-2h-inden-5-ol, 4,6-bis methylethyl-1,1,3,3-tetramethylindan-5-ol, 1,1,3,3-tetramethyl-4,6-di propan-2-yl-2h-inden-5-ol, 1h-inden-5-ol, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl-1h-inden-5-ol
PubChem CID 621162
IUPAC Name 1,1,3,3-tetramethyl-4,6-di(propan-2-yl)-2H-inden-5-ol
SMILES CC(C)C1=C(C(=C2C(=C1)C(CC2(C)C)(C)C)C(C)C)O

Specifications

Melting Point 146°C to 148°C
Quantity 1 g
Formula Weight 274.45
Percent Purity 97%
Chemical Name or Material 4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'