missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-1,3,5-triazin-2-amine hydrate, ≥95%, Thermo Scientific™

Product Code. 10334392
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-59-8
Molecular Formula C5H10N4O
Molecular Weight (g/mol) 142.16
MDL Number MFCD00663061
InChI Key CZKUOCRCKNMNTI-UHFFFAOYSA-N
Synonym 4,6-dimethyl-1,3,5-triazin-2-amine hydrate, dimethyl-1,3,5-triazin-2-amine hydrate, 1,3,5-triazin-2-amine,4,6-dimethyl-, hydrate 1:1, 2-amino-4,6-dimethyl-1,3,5-triazine hydrate, 4,6-dimethyl-1,3,5-triazine-2-ylamine, oxamethane, 1,3,5-triazin-2-amine,4,6-dimethyl-,hydrate 1:1
PubChem CID 2782054
IUPAC Name 4,6-dimethyl-1,3,5-triazin-2-amine;hydrate
SMILES O.CC1=NC(N)=NC(C)=N1

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 142.17
Percent Purity 95%
Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'