missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Hydrazino-4-pyrimidinyl)morpholine, 96%, Thermo Scientific™

Product Code. 15476427
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5767-36-2
Molecular Formula C8H13N5O
Molecular Weight (g/mol) 195.226
MDL Number MFCD11655642
InChI Key XWDNLTCJJYYYCI-UHFFFAOYSA-N
Synonym 4-6-hydrazinylpyrimidin-4-yl morpholine, 4-6-hydrazinopyrimidin-4-yl morpholine, morpholine, 4-6-hydrazinyl-4-pyrimidinyl, 4-6-hydrazino-4-pyrimidinyl morpholine, 6-morpholin-4-ylpyrimidine-4-ylhydrazine, 4-6-hydrazino-pyrimidin-4-yl-morpholine, 6-morpholin-4-yl-pyrimidin-4-yl-hydrazine
PubChem CID 13393622
IUPAC Name (6-morpholin-4-ylpyrimidin-4-yl)hydrazine
SMILES C1COCCN1C2=CC(=NC=N2)NN

Specifications

Melting Point 162°C to 163°C
Quantity 1 g
Formula Weight 195.22
Percent Purity 96%
Chemical Name or Material 4-(6-Hydrazino-4-pyrimidinyl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.