missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Methyl-2-benzimidazolyl)benzamidoxime, 97%, Thermo Scientific™

Product Code. 15488857
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256486-22-2
Molecular Formula C15H14N4O
Molecular Weight (g/mol) 266.304
MDL Number MFCD16658868
InChI Key SQIGGAWUCHCZKN-UHFFFAOYSA-N
Synonym 4-6-methyl-2-benzimidazolyl benzamidoxime, afqxausvhhhsmn-uhfffaoysa-n, n-hydroxy-4-6-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide, n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide
PubChem CID 73995333
IUPAC Name [4-(5-methyl-1,3-dihydrobenzimidazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-nitrosomethanamine
SMILES CC1=CC2=C(C=C1)NC(=C3C=CC(=C(N)N=O)C=C3)N2

Specifications

Quantity 5 g
Formula Weight 266.3
Percent Purity 97%
Chemical Name or Material 4-(6-Methyl-2-benzimidazolyl)benzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'