missing translation for 'onlineSavingsMsg'
Learn More

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%, Thermo Scientific Chemicals

78.40 € - 618.00 €

Chemical Identifiers

CAS 31364-42-8
Molecular Formula C16H32N2O5
Molecular Weight (g/mol) 332.44
MDL Number MFCD00005108
InChI Key HDLXPNDSLDLJHF-UHFFFAOYSA-N
Synonym 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221
PubChem CID 123438
IUPAC Name 4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
SMILES C1COCCN2CCOCCOCCN(CCO1)CCOCC2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11475220
View Documents
Thermo Scientific Alfa Aesar
044504.77
0.1 g
78.40 €
0.10g
Please to purchase this item. Need a web account? Register with us today!
11455220
View Documents
Thermo Scientific Alfa Aesar
044504.02
0.5 g
230.00 €
0.50g
Please to purchase this item. Need a web account? Register with us today!
11465220
View Documents
Thermo Scientific Alfa Aesar
044504.04
2 g
618.00 €
2g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is used as a phase transfer catalysts by transferring ions from one phase to another. It is involved in the synthesis of alkalides and electrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31364-42-8
332.44
HDLXPNDSLDLJHF-UHFFFAOYSA-N
123438
C1COCCN2CCOCCOCCN(CCO1)CCOCC2
C16H32N2O5
MFCD00005108
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane

Specifications

31364-42-8
1.11 g/mL
1.501
616444
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221
HDLXPNDSLDLJHF-UHFFFAOYSA-N
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
123438
97%
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
Yellow
C16H32N2O5
MFCD00005108
Light sensitive
Miscible with water.
C1COCCN2CCOCCOCCN(CCO1)CCOCC2
332.44
332.44
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 250-592-1

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only