missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals

18.89 € - 98.29 €

Chemical Identifiers

CAS 86-98-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.05
MDL Number MFCD00006774
InChI Key HXEWMTXDBOQQKO-UHFFFAOYSA-N
PubChem CID 6866
IUPAC Name 4,7-dichloroquinoline
SMILES C1=CC2=C(C=CN=C2C=C1Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10106033
View Documents
Thermo Scientific Acros
113750050
5 g Glass bottle
18.89 €
5g
Please to purchase this item. Need a web account? Register with us today!
10305990
View Documents
Thermo Scientific
113750250
25 g Glass bottle
35.38 €
25g
Please to purchase this item. Need a web account? Register with us today!
15780789
View Documents
Thermo Scientific Acros
113751000
100 g Glass bottle
98.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86-98-6
198.05
HXEWMTXDBOQQKO-UHFFFAOYSA-N
4,7-dichloroquinoline
C9H5Cl2N
MFCD00006774
6866
C1=CC2=C(C=CN=C2C=C1Cl)Cl

Specifications

86-98-6
100.0
Brown to White
164°C
98.5% min. (GC)
MFCD00006774
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid,174g/L toluene ( 20 c),soluble in acetone,methanol and ethanol
C1=CC2=C(C=CN=C2C=C1Cl)Cl
198.05
198.05
Powder
97.5
84°C to 86°C
148°C
Authentic
C9H5Cl2N
quinoline, 4,7-dichloro, 4,7-dichloro-quinoline, unii-z61o2heq2j, z61o2heq2j, 4,7dichloroquinoline, pubchem5880, 4.7-dichloroquinoline, 4,7 dichloroquinoline, quinoline,7-dichloro, amodiaquine impurity 1
HXEWMTXDBOQQKO-UHFFFAOYSA-N
4,7-dichloroquinoline
6866
98%
4, 7-Dichloroquinoline, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 201-714-7

missing translation for 'rtecsNumber' : VB4200000

missing translation for 'tsca' : TSCA

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.