missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetamido-2'-methylacetophenone, 97%, Thermo Scientific Chemicals

73.50 € - 229.00 €

Chemical Identifiers

CAS 34956-31-5
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD02258876
InChI Key PTARWPNYVATTDE-UHFFFAOYSA-N
Synonym n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl
PubChem CID 2737381
IUPAC Name N-(4-acetyl-3-methylphenyl)acetamide
SMILES CC(=O)NC1=CC(C)=C(C=C1)C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11431520
View Documents
Thermo Scientific Alfa Aesar
H26933.06
5 g
73.50 €
5g
Please to purchase this item. Need a web account? Register with us today!
11441520
View Documents
Thermo Scientific Alfa Aesar
H26933.14
25 g
229.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34956-31-5
191.23
PTARWPNYVATTDE-UHFFFAOYSA-N
2737381
CC(=O)NC1=CC(C)=C(C=C1)C(C)=O
C11H13NO2
MFCD02258876
n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl
N-(4-acetyl-3-methylphenyl)acetamide

Specifications

34956-31-5
C11H13NO2
n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl
CC(=O)NC1=CC(C)=C(C=C1)C(C)=O
191.23
191.23
4'-Acetamido-2'-methylacetophenone
137°C to 140°C
MFCD02258876
PTARWPNYVATTDE-UHFFFAOYSA-N
N-(4-acetyl-3-methylphenyl)acetamide
2737381
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only