missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals

Product Code. 11496320
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103204-69-9
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD02258874
InChI Key AQPDTYYKDYMCTH-UHFFFAOYSA-N
Synonym 4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid
PubChem CID 2735224
IUPAC Name 4-acetamido-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)NC(=O)C)C(=O)O

Specifications

Melting Point ∽248°C (decomposition)
Quantity 25 g
Formula Weight 193.21
Percent Purity 96%
Chemical Name or Material 4-Acetamido-2-methylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'