missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidocinnamic acid, predominantly trans, 98%, Thermo Scientific Chemicals

Product Code. 11377375
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918352
MDL Number MFCD00016846

Specifications

Melting Point ∼259°C (decomposition)
Molecular Formula C11H11NO3
Quantity 2 g
Beilstein 2724869
Synonym 3-4-acetylamino phenyl acrylic acid, 4-acetylaminocinnamic acid, 4-acetamidocinnamic acid, 2e-3-4-acetamidophenyl prop-2-enoic acid, 3-4-acetamidophenyl acrylic acid, 2e-3-4-acetylamino phenyl-2-propenoic acid, e-3-4-acetamidophenyl acrylic acid, 2e-3-4-acetylamino phenyl acrylic acid, e-3-4-acetamidophenyl prop-2-enoic acid, 2e-3-4-acetylamino phenyl prop-2-enoic acid
InChI Key WGMFHSADKZJPGR-QPJJXVBHSA-N
SMILES CC(=O)NC1=CC=C(C=C1)C=CC(=O)O
IUPAC Name (E)-3-(4-acetamidophenyl)prop-2-enoic acid
Molecular Weight (g/mol) 205.21
PubChem CID 5357155
Formula Weight 205.21
Percent Purity 0.98
Chemical Name or Material 4-Acetamidocinnamic acid, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.