missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidocinnamic acid, predominantly trans, 98%, Thermo Scientific Chemicals

Product Code. 11387375
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918352
MDL Number MFCD00016846
ChEBI CHEBI:16388

Specifications

Melting Point ∼259°C (decomposition)
Molecular Formula C8H8O4
Quantity 10 g
Beilstein 2724869
Synonym 2-hydroxy-2-4-hydroxyphenyl acetic acid hydrate, 4-hydroxymandelic acid monohydrate, dl-4-hydroxymandelic acid monohydrate, 4-hydroxymandelic acid hydrate, 3-hydroxymandelic acid monohydrate, hydroxy 4-hydroxyphenyl acetic acid-water 1/1, hydroxy-4-hydroxy-phenyl-acetic acid monohydrate, benzeneacetic acid, a,4-dihydroxy-, hydrate 1:1
InChI Key YHXHKYRQLYQUIH-UHFFFAOYNA-N
SMILES C1=CC(=CC=C1C(C(=O)O)O)O
IUPAC Name 2-hydroxy-2-(4-hydroxyphenyl)acetic acid
Molecular Weight (g/mol) 168.15
PubChem CID 328
Formula Weight 205.21
Percent Purity 0.98
Chemical Name or Material 4-Acetamidocinnamic acid, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.