missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidothiophenol, 95%, Thermo Scientific™

57.00 €

Chemical Identifiers

CAS 1126-81-4
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.23
MDL Number MFCD00004848
InChI Key AYEQJLOHMLYKAV-UHFFFAOYSA-N
Synonym 4-acetamidothiophenol, 4'-mercaptoacetanilide, 4-acetamidobenzenethiol, n-4-sulfanylphenyl acetamide, 4-acetamido thiophenol, n-4-mercaptophenyl acetamide, acetanilide, 4'-mercapto, 4-acetaminothiophenol, 4-mercaptoacetanilide, 4-acetylaminothiophenol
PubChem CID 70777
IUPAC Name N-(4-sulfanylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(S)C=C1
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10152890
View Documents
Thermo Scientific Acros
102340050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1126-81-4
167.23
AYEQJLOHMLYKAV-UHFFFAOYSA-N
70777
CC(=O)NC1=CC=C(S)C=C1
C8H9NOS
MFCD00004848
4-acetamidothiophenol, 4'-mercaptoacetanilide, 4-acetamidobenzenethiol, n-4-sulfanylphenyl acetamide, 4-acetamido thiophenol, n-4-mercaptophenyl acetamide, acetanilide, 4'-mercapto, 4-acetaminothiophenol, 4-mercaptoacetanilide, 4-acetylaminothiophenol
N-(4-sulfanylphenyl)acetamide

Specifications

1126-81-4
100.0
Yellow
95%
CH3CONHC6H4SH
13, 541
AYEQJLOHMLYKAV-UHFFFAOYSA-N
N-(4-sulfanylphenyl)acetamide
70777
95%
4-Acetamidothiophenol, 95%
94.0
149°C to 154°C
Authentic
C8H9NOS
MFCD00004848
4-acetamidothiophenol, 4'-mercaptoacetanilide, 4-acetamidobenzenethiol, n-4-sulfanylphenyl acetamide, 4-acetamido thiophenol, n-4-mercaptophenyl acetamide, acetanilide, 4'-mercapto, 4-acetaminothiophenol, 4-mercaptoacetanilide, 4-acetylaminothiophenol
CC(=O)NC1=CC=C(S)C=C1
167.23
167.23
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-427-7

Product Identifier
  • 4-Acetamidothiophenol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only