missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-3-methoxycinnamic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11444163
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2596-47-6
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00014378
InChI Key IHKNVZISLLDMOR-GQCTYLIASA-N
Synonym acetylferulic acid, 4-acetoxy-3-methoxycinnamic acid, 3-methoxy-4-acetoxycinnamic acid, 3-4-acetoxy-3-methoxyphenyl acrylic acid, cinnamic acid, 4-acetoxy-3-methoxy, acetylated ferulic acid, cinnamic acid, 4-hydroxy-3-methoxy-, acetate, 2-propenoic acid, 3-4-acetyloxy-3-methoxyphenyl, 2e-3-4-acetyloxy-3-methoxyphenyl prop-2-enoic acid
PubChem CID 5354677
ChEBI CHEBI:86582
IUPAC Name (E)-3-(4-acetyloxy-3-methoxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=C(C=C(C=C1)C=CC(=O)O)OC

Specifications

Melting Point 193°C to 196°C
Odor Odorless
Quantity 1 g
Beilstein 2218026
Formula Weight 236.22
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxy-3-methoxycinnamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'