missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetoxyacetophenone, 99%, Thermo Scientific Chemicals

25.40 € - 252.00 €

Chemical Identifiers

CAS 13031-43-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017229
InChI Key SMIOEQSLJNNKQF-UHFFFAOYSA-N
Synonym 4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
PubChem CID 83063
ChEBI CHEBI:86558
IUPAC Name (4-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11425956
View Documents
Thermo Scientific Alfa Aesar
B22284.14
25 g
25.40 €
25g
Please to purchase this item. Need a web account? Register with us today!
11435956
View Documents
Thermo Scientific Alfa Aesar
B22284.22
100 g
68.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
11445956
View Documents
Thermo Scientific Alfa Aesar
B22284.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

Solubility
Slightly soluble in water(4450 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

13031-43-1
178.19
SMIOEQSLJNNKQF-UHFFFAOYSA-N
83063
(4-acetylphenyl) acetate
C10H10O3
MFCD00017229
4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
CHEBI:86558
CC(=O)OC1=CC=C(C=C1)C(C)=O

Specifications

13031-43-1
165°C to 170°C (14 mmHg)
MFCD00017229
4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
SMIOEQSLJNNKQF-UHFFFAOYSA-N
(4-acetylphenyl) acetate
83063
178.19
4'-Acetoxyacetophenone
51°C to 54°C
C10H10O3
2093080
Slightly soluble in water(4450mg/L).
CC(=O)OC1=CC=C(C=C1)C(C)=O
178.19
CHEBI:86558
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-894-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.