missing translation for 'onlineSavingsMsg'
Learn More
4-Acetoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals Product Code.: 15449657

4-Acetoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15449657
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15449657

Brand: Thermo Scientific Alfa Aesar H52879.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177490-82-3
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD09027198
InChI Key VILXJXDXZGKJLU-UHFFFAOYSA-N
Synonym 4-acetoxyphenyl boronic acid, 4-acetoxyphenylboronic acid, 4-acetoxybenzeneboronic acid, 4-acetyloxy phenylboronic acid, boronic acid, 4-acetyloxy phenyl, boronic acid,b-4-acetyloxy phenyl, 4-boronophenyl acetate, pubchem20170, acmc-209ecy, 4-acetoxyphenyl boronicacid
PubChem CID 44119577
IUPAC Name (4-acetyloxyphenyl)boronic acid
SMILES CC(=O)OC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 220°C to 224°C
Quantity 1 g
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 4-Acetoxybenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX04H054 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52879.03
Lot Number10170231 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52879.03
Lot Number81202405 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52879.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Acetoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.