missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-N-methylaniline, 97%, Thermo Scientific Chemicals

37.99 € - 100.00 €

Chemical Identifiers

CAS 17687-47-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD09261160
InChI Key GDBAYICEBDZUNM-UHFFFAOYSA-N
Synonym 4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone
PubChem CID 11607936
IUPAC Name 1-[4-(methylamino)phenyl]ethanone
SMILES CNC1=CC=C(C=C1)C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431518
SDS View Documents
Thermo Scientific Alfa Aesar
H55301.MD
250 mg
37.99 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15411518
SDS View Documents
Thermo Scientific Alfa Aesar
H55301.03
1 g
100.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17687-47-7
149.19
GDBAYICEBDZUNM-UHFFFAOYSA-N
11607936
CNC1=CC=C(C=C1)C(C)=O
C9H11NO
MFCD09261160
4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone
1-[4-(methylamino)phenyl]ethanone

Specifications

17687-47-7
Brown to White
MFCD09261160
GDBAYICEBDZUNM-UHFFFAOYSA-N
1-[4-(methylamino)phenyl]ethanone
11607936
97%
4-Acetyl-N-methylaniline
103°C to 107°C
C9H11NO
4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone
CNC1=CC=C(C=C1)C(C)=O
149.19
149.19
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P285-P304+P341-P342+P311-P501c

H334

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.