Learn More
4-Acetylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
14.37 € - 274.37 €
Chemical Identifiers
CAS | 1788-10-9 |
---|---|
Molecular Formula | C8H7ClO3S |
Molecular Weight (g/mol) | 218.65 |
MDL Number | MFCD00800269 |
InChI Key | FXVDNCRTKXMSEZ-UHFFFAOYSA-N |
Synonym | 4-acetylbenzene-1-sulfonyl chloride, 4-acetyl-benzenesulfonyl chloride, 4-acetylphenylsulfonyl chloride, benzenesulfonylchloride, 4-acetyl, benzenesulfonyl chloride, 4-acetyl, 1-acetyl-4-chlorosulfonyl benzene, 4-acetylbenzenesulfonyl chloride, acmc-1bs75, ksc183k3b, 4-acetylbenzenesulfonylchloride |
PubChem CID | 266418 |
IUPAC Name | 4-acetylbenzenesulfonyl chloride |
SMILES | CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10479242
|
Thermo Scientific Acros
343850010 |
1 g | Glass bottle | N/A | |||||
10387651
|
Thermo Scientific Acros
343850050 |
5 g | Glass bottle |
51.46 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10064103
|
Thermo Scientific Acros
343850500 |
50 g | Glass bottle |
274.37 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1788-10-9 | |
218.65 | |
FXVDNCRTKXMSEZ-UHFFFAOYSA-N | |
266418 | |
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl |
C8H7ClO3S | |
MFCD00800269 | |
4-acetylbenzene-1-sulfonyl chloride, 4-acetyl-benzenesulfonyl chloride, 4-acetylphenylsulfonyl chloride, benzenesulfonylchloride, 4-acetyl, benzenesulfonyl chloride, 4-acetyl, 1-acetyl-4-chlorosulfonyl benzene, 4-acetylbenzenesulfonyl chloride, acmc-1bs75, ksc183k3b, 4-acetylbenzenesulfonylchloride | |
4-acetylbenzenesulfonyl chloride |
Specifications
1788-10-9 | |
100.0 | |
Brown to White | |
96% min. (GC) | |
H3CCO(C6H4)SO2Cl | |
4-acetylbenzene-1-sulfonyl chloride, 4-acetyl-benzenesulfonyl chloride, 4-acetylphenylsulfonyl chloride, benzenesulfonylchloride, 4-acetyl, benzenesulfonyl chloride, 4-acetyl, 1-acetyl-4-chlorosulfonyl benzene, 4-acetylbenzenesulfonyl chloride, acmc-1bs75, ksc183k3b, 4-acetylbenzenesulfonylchloride | |
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl | |
218.65 | |
218.65 | |
Chunks and/or Powder |
96.0 | |
83°C to 87°C | |
Authentic | |
C8H7ClO3S | |
MFCD00800269 | |
FXVDNCRTKXMSEZ-UHFFFAOYSA-N | |
4-acetylbenzenesulfonyl chloride | |
266418 | |
97% | |
4-Acetylbenzenesulfonyl chloride, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
- 4-Acetylbenzenesulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.