missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
66.23 € - 273.36 €
Chemical Identifiers
CAS | 171364-81-1 |
---|---|
Molecular Formula | C14H19BO3 |
Molecular Weight (g/mol) | 246.11 |
MDL Number | MFCD05863922 |
InChI Key | BATKIZWNRQGSKE-UHFFFAOYSA-N |
Synonym | 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone |
PubChem CID | 2760596 |
IUPAC Name | 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone |
SMILES | CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11956031
|
Thermo Scientific Acros
444430010 |
1 g |
66.23 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11966031
|
Thermo Scientific Acros
444430050 |
5 g |
273.36 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
171364-81-1 | |
246.11 | |
BATKIZWNRQGSKE-UHFFFAOYSA-N | |
2760596 | |
CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
C14H19BO3 | |
MFCD05863922 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone |
Specifications
171364-81-1 | |
White | |
Authentic | |
Glass Bottle | |
MFCD05863922 | |
BATKIZWNRQGSKE-UHFFFAOYSA-N | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone | |
2760596 | |
97% | |
4-Acetylphenylboronic acid pinacol ester |
59°C to 62°C | |
80°C (0.1 Torr) | |
97% | |
C14H19BO3 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone | |
CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
246.11 | |
246.12 | |
Crystalline Powder |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Acetylphenylboronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title