missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride, Thermo Scientific™

Product Code. 15469205
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301739-00-3
Molecular Formula C11H18ClN3O
Molecular Weight (g/mol) 243.735
MDL Number MFCD11869912
InChI Key QBVBNVDHWTXFAT-UHFFFAOYSA-N
Synonym 4-amino-1-3-methoxy-2-pyridyl piperidine hydrochloride, 1-3-methoxypyridin-2-yl piperidin-4-amine hydrochloride, 2-4-amino-1-piperidinyl-3-methoxypyridine, 1-3-methoxy-2-pyridyl-4-piperidinamine hydrochloride
PubChem CID 53400829
IUPAC Name 1-(3-methoxypyridin-2-yl)piperidin-4-amine;hydrochloride
SMILES COC1=C(N=CC=C1)N2CCC(CC2)N.Cl

Specifications

Quantity 1 g
Formula Weight 243.73
Chemical Name or Material 4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'