missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-butanol, 98%, Thermo Scientific Chemicals

27.84 € - 335.67 €

Chemical Identifiers

CAS 13325-10-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008230
InChI Key BLFRQYKZFKYQLO-UHFFFAOYSA-N
Synonym 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
PubChem CID 25868
IUPAC Name 4-aminobutan-1-ol
SMILES C(CCO)CN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470563
View Documents
Thermo Scientific Alfa Aesar
A12680.03
1 g
27.84 €
1g
Please to purchase this item. Need a web account? Register with us today!
11480563
View Documents
Thermo Scientific Alfa Aesar
A12680.06
5 g
79.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11490563
View Documents
Thermo Scientific Alfa Aesar
A12680.14
25 g
335.67 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

Solubility
Miscible with water.

Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.
Specifications

Chemical Identifiers

13325-10-5
89.138
BLFRQYKZFKYQLO-UHFFFAOYSA-N
25868
C(CCO)CN
C4H11NO
MFCD00008230
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
4-aminobutan-1-ol

Specifications

13325-10-5
0.967
107°C (224°F)
C4H11NO
1.461
UN2735
Air sensitive; Hygroscopic
Miscible with water.
C(CCO)CN
89.138
89.14
4-Amino-1-butanol
16°C to 18°C
206°C
Amine-like
H2N(CH2)4OH
MFCD00008230
1731411
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
BLFRQYKZFKYQLO-UHFFFAOYSA-N
4-aminobutan-1-ol
25868
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 236-364-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.