missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-difluorobenzonitrile, 96%, Thermo Scientific Chemicals

Product Code. 11455637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112279-61-5
Molecular Formula C7H4F2N2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00190102
InChI Key LAPGMTOHOQPDGI-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75
PubChem CID 2735900
IUPAC Name 4-amino-2,5-difluorobenzonitrile
SMILES C1=C(C(=CC(=C1F)N)F)C#N

Specifications

Melting Point 91°C to 94°C
Quantity 1 g
UN Number UN3439
Formula Weight 154.12
Percent Purity 96%
Chemical Name or Material 4-Amino-2,5-difluorobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'