missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorophenol, 97%, Thermo Scientific Chemicals

71.70 € - 277.00 €

Chemical Identifiers

CAS 399-96-2
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00671760
InChI Key MXJQJURZHQZLNN-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyaniline, 2-fluoro-4-aminophenol, phenol, 4-amino-2-fluoro, 4-amino-2-fluoro-phenol, zlchem 955, pubchem2831, acmc-1cswx, 4-amino-2-fluoro phenol
PubChem CID 2735918
IUPAC Name 4-amino-2-fluorophenol
SMILES C1=CC(=C(C=C1N)F)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435700
View Documents
Thermo Scientific Alfa Aesar
H25936.03
1 g
71.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11445700
View Documents
Thermo Scientific Alfa Aesar
H25936.06
5 g
277.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

399-96-2
127.118
MXJQJURZHQZLNN-UHFFFAOYSA-N
2735918
C1=CC(=C(C=C1N)F)O
C6H6FNO
MFCD00671760
3-fluoro-4-hydroxyaniline, 2-fluoro-4-aminophenol, phenol, 4-amino-2-fluoro, 4-amino-2-fluoro-phenol, zlchem 955, pubchem2831, acmc-1cswx, 4-amino-2-fluoro phenol
4-amino-2-fluorophenol

Specifications

399-96-2
C6H6FNO
3-fluoro-4-hydroxyaniline, 2-fluoro-4-aminophenol, phenol, 4-amino-2-fluoro, 4-amino-2-fluoro-phenol, zlchem 955, pubchem2831, acmc-1cswx, 4-amino-2-fluoro phenol
C1=CC(=C(C=C1N)F)O
127.118
127.12
4-Amino-2-fluorophenol
170°C to 172°C
MFCD00671760
MXJQJURZHQZLNN-UHFFFAOYSA-N
4-amino-2-fluorophenol
2735918
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.