Learn More
4-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
57.70 € - 165.00 €
Chemical Identifiers
CAS | 819058-34-9 |
---|---|
Molecular Formula | C12H17BFNO2 |
Molecular Weight (g/mol) | 237.08 |
MDL Number | MFCD09033884 |
InChI Key | AIXGNRNTXUKZLC-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 17906172 |
IUPAC Name | 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
SMILES | CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479745
|
Thermo Scientific Alfa Aesar
H52349.MD |
250 mg |
57.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15459745
|
Thermo Scientific Alfa Aesar
H52349.03 |
1 g |
165.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
819058-34-9 | |
237.08 | |
AIXGNRNTXUKZLC-UHFFFAOYSA-N | |
17906172 | |
CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1 |
C12H17BFNO2 | |
MFCD09033884 | |
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
819058-34-9 | |
MFCD09033884 | |
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1 | |
237.08 | |
237.08 | |
4-Amino-3-fluorobenzeneboronic acid pinacol ester |
C12H17BFNO2 | |
Air Sensitive | |
AIXGNRNTXUKZLC-UHFFFAOYSA-N | |
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
17906172 | |
96% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.