missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-nitrodiphenyl sulfide, Thermo Scientific™

Product Code. 17565525
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 101-59-7
Molecular Formula C12H10N2O2S
Molecular Weight (g/mol) 246.28
MDL Number MFCD00007881
InChI Key ZBPKGHOGUVVDLF-UHFFFAOYSA-N
IUPAC Name 4-[(4-nitrophenyl)sulfanyl]aniline
SMILES NC1=CC=C(SC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Alpha Vector AMINONITRODIPHENYLSULFIDE
Melting Point 141°C to 145°C
Color Light yellow to brown
Infrared Spectrum Conforms
Assay Percent Range ≥97.5% (Titration with HClO4)
Packaging Glass Bottle
Linear Formula H2NC6H4SC6H4NO2
Quantity 1 g
Beilstein 13-534
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4'-nitrodiphenyl sulfide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'