missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 15426867
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14160-93-1
Molecular Formula C5H4ClN3O
Molecular Weight (g/mol) 157.56
MDL Number MFCD03789626
InChI Key GOJNFUXEBVBARW-UHFFFAOYSA-N
Synonym 4-amino-6-chloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 4-amino-6-chloro, 4-amino-6-chloro-5-pyrimidinecarbaldehyde, 4-amino-6-chloro-pyrimidine-5-carbaldehyde, 4-amino-6-chloro-5-pyrimidinecarboxaldehyde, 4-amino-6-chloro-5-formylpyrimidine, 4-amino-6-chloro-5-formyl-1,3-diazine, pubchem22541, 6-amino-4-chloropyrimidine-5-carbaldehyde, 4-amino-6-chloro-pyrimidine-5-carboxaldehyde
PubChem CID 84230
IUPAC Name 4-amino-6-chloropyrimidine-5-carbaldehyde
SMILES NC1=NC=NC(Cl)=C1C=O

Specifications

Melting Point >300°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 4-Amino-6-chloropyrimidine-5-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'