missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11318965
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2498-50-2
Molecular Formula C7H11Cl2N3
Molecular Weight (g/mol) 208.086
MDL Number MFCD00013001
InChI Key GHEHNICLPWTXJC-UHFFFAOYSA-N
Synonym 4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
PubChem CID 75626
ChEBI CHEBI:10618
IUPAC Name 4-aminobenzenecarboximidamide;dihydrochloride
SMILES C1=CC(=CC=C1C(=N)N)N.Cl.Cl

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 3692927
Sensitivity Hygroscopic
Formula Weight 208.09
Percent Purity 97%
Chemical Name or Material 4-Aminobenzamidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.