missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzo-2,1,3-thiadiazole, 98%, Thermo Scientific Chemicals

Product Code. 11312207
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 767-64-6
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.187
MDL Number MFCD00005810
InChI Key DRLGIZIAMHIQHL-UHFFFAOYSA-N
Synonym 4-aminobenzo-2,1,3-thiadiazole, 4-amino-2,1,3-benzothiadiazole, 4-aminopiazthiole, benzo c 1,2,5 thiadiazol-4-amine, 2,1,3-benzothiadiazole, 4-amino, 7-amino-2,1,3-benzothiadiazole, benzo 1,2,5 thiadiazol-4-ylamine, 2,1,3-benzothiadiazole, 7-amino, 2,1,3-benzothiadiazol-4-ylamine, 2,1,3-benzothiadiazol-4 1h-imine
PubChem CID 69845
IUPAC Name 2,1,3-benzothiadiazol-4-amine
SMILES C1=CC2=NSN=C2C(=C1)N

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
Beilstein 3551
Solubility Information Insoluble in water.
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 4-Aminobenzo-2,1,3-thiadiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.