missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 15425679
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmaceutical intermediate

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-06-6
Molecular Formula C7H6NNaO2
Molecular Weight (g/mol) 159.12
MDL Number MFCD00064395
InChI Key XETSAYZRDCRPJY-UHFFFAOYSA-M
Synonym sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
PubChem CID 517441
IUPAC Name sodium;4-aminobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Color Yellow
Quantity 100 g
Solubility Information Soluble in water (50mg/ml).
Formula Weight 159.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminobenzoic acid sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'