missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.
Solubility
Soluble in alcohol, ether and benzene. Partially soluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents and strong acids. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.
Solubility
Soluble in alcohol, ether and benzene. Partially soluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents and strong acids. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Melting Point | 59°C to 63°C |
Quantity | 5 g |
Beilstein | 2078680 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in alcohol,ether and benzene. Partially soluble in water. |
Formula Weight | 123.16 |
Percent Purity | 98% |
Chemical Name or Material | 4-Aminobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'