missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzyl cyanide, 99%, Thermo Scientific Chemicals

Product Code. 10010560
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3544-25-0
CAS Min % 98.5
CAS Max % 100.0
Melting Point 42.0°C to 46.0°C
Color Yellow to Brown
Boiling Point 312.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8N2
Linear Formula H2NC6H4CH2CN
MDL Number MFCD00007912
Quantity 10 g
Beilstein 14, 457
Synonym 4-aminobenzyl cyanide, 4-aminophenylacetonitrile, 2-4-aminophenyl acetonitrile, 4-aminophenyl acetonitrile, p-aminobenzyl cyanide, benzeneacetonitrile, 4-amino, 4-aminobenzylcyanide, 4-amino-phenyl-acetonitrile, 2-4-aminophenyl ethanenitrile, 4-aminobenzyl
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,organic solvents,slightly soluble in cs2
InChI Key YCWRFIYBUQBHJI-UHFFFAOYSA-N
SMILES NC1=CC=C(CC#N)C=C1
IUPAC Name 2-(4-aminophenyl)acetonitrile
Molecular Weight (g/mol) 132.17
PubChem CID 77000
Formula Weight 132.16
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Aminobenzyl cyanide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminobenzyl cyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.