missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminomethyltetrahydropyran, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.02g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 115.17 |
Percent Purity | 97% |
Chemical Name or Material | 1-Tetrahydro-2H-pyran-4-ylmethanamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction