missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomorpholine, 95%, Thermo Scientific Chemicals

65.43 € - 575.87 €

Chemical Identifiers

CAS 4319-49-7
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00006174
InChI Key MKQLBNJQQZRQJU-UHFFFAOYSA-N
Synonym 4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
PubChem CID 20315
IUPAC Name morpholin-4-amine
SMILES C1COCCN1N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469327
View Documents
Thermo Scientific Alfa Aesar
B21381.06
5 g
65.43 €
5g
Please to purchase this item. Need a web account? Register with us today!
11479327
View Documents
Thermo Scientific Alfa Aesar
B21381.14
25 g
209.04 €
25g
Please to purchase this item. Need a web account? Register with us today!
11489327
View Documents
Thermo Scientific Alfa Aesar
B21381.22
100 g
575.87 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4319-49-7
102.137
MKQLBNJQQZRQJU-UHFFFAOYSA-N
20315
C1COCCN1N
C4H10N2O
MFCD00006174
4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
morpholin-4-amine

Specifications

4319-49-7
168°C
C4H10N2O
MFCD00006174
Air sensitive
MKQLBNJQQZRQJU-UHFFFAOYSA-N
morpholin-4-amine
20315
98%
1.059
58°C (137°F)
1.4785
UN2733
4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
C1COCCN1N
102.137
102.14
4-Aminomorpholine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 224-347-4

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.