missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminopropiophenone, 98%, Thermo Scientific Chemicals

Product Code. 11965321
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-69-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00017113
InChI Key FSWXOANXOQPCFF-UHFFFAOYSA-N
Synonym 4'-aminopropiophenone, p-aminopropiophenone, 1-4-aminophenyl propan-1-one, 1-4-aminophenyl-1-propanone, 1-propanone, 1-4-aminophenyl, 4-aminopropiophenone, propiophenone, 4'-amino, usaf uctl-1856, ethyl p-aminophenyl ketone, paraminopropiophenone
PubChem CID 6270
IUPAC Name 1-(4-aminophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 135.0°C to 140.0°C
Assay Percent Range 98%
Linear Formula NH2C6H4C(O)CH2CH3
Quantity 50 g
Merck Index 15,466
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, insoluble in diethyl ether
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 4'-Aminopropiophenone
compliance-icons
Product Identifier
  • 4'-Aminopropiophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.