missing translation for 'onlineSavingsMsg'
Learn More

4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals

Product Code. 10172700
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-49-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007789
InChI Key WUBBRNOQWQTFEX-UHFFFAOYSA-N
Synonym 4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
PubChem CID 4649
ChEBI CHEBI:27565
IUPAC Name 4-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 147°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H3(OH)CO2H
Quantity 5 g
Beilstein 14, 579
Merck Index 15, 473
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in dilute hno3 and dilute naoh,aceton,1g/21mL alcohol,slightly soluble in ether,practically insoluble in benzene,chloroform,,carbontetrachloride
Formula Weight 153.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Aminosalicylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.